Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDCCAG1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SDCCAG1.
Supplier: Abnova Corporation PAB17341
Specifications
SDCCAG1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of SDCCAG1. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
FLJ10051/NY-CO-1 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total SDCCAG1 protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
SDCCAG1 | |
SDCCAG1 | |
A synthetic peptide corresponding to internal of human SDCCAG1. | |
RUO | |
9147 | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction