Learn More
SELE Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SELE.
Supplier: Abnova Corporation PAB14854
Description
The protein encoded by this gene is found in cytokine-stimulated endothelial cells and is thought to be responsible for the accumulation of blood leukocytes at sites of inflammation by mediating the adhesion of cells to the vascular lining. It exhibits structural features such as the presence of lectin- and EGF-like domains followed by short consensus repeat (SCR) domains that contain 6 conserved cysteine residues. These proteins are part of the selectin family of cell adhesion molecules. Adhesion molecules participate in the interaction between leukocytes and the endothelium and appear to be involved in the pathogenesis of atherosclerosis. [provided by RefSeq
Specifications
| SELE | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SELE. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| CD62E/ELAM/ELAM1/ESEL/LECAM2 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 6401 | |
| Bovine, Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| SELE | |
| SELE | |
| A synthetic peptide corresponding to N-terminus of human SELE. | |
| 100 μg | |
| Primary | |
| This antibody recognizes ∼65 KDa of SELE. | |
| Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only