Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEMA4D Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SEMA4D.
Supplier: Abnova Corporation PAB0990
Description
Sequence: KPKSDFCDREQSSpecifications
SEMA4D | |
Polyclonal | |
sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D | |
SEMA4D | |
SEMA4D | |
A synthetic peptide corresponding to amino acids 777-788 of human SEMA4D. | |
RUO | |
10507 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
In PBS (0.02% sodium azide) | |
C9orf164, CD100, FLJ33485, FLJ34282, FLJ39737, FLJ46484, M-sema-G, MGC169138, MGC169141, SEMAJ, coll-4 | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction