Learn More
SENP2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SENP2.
Supplier: Abnova Corporation PAB2415
Description
SUMO1 (UBL1; MIM 601912) is a small ubiquitin-like protein that can be covalently conjugated to other proteins. SENP2 is one of a group of enzymes that process newly synthesized SUMO1 into the conjugatable form and catalyze the deconjugation of SUMO1-containing species.[supplied by OMIM
Specifications
SENP2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of SENP2. | |
In PBS (0.09% sodium azide) | |
AXAM2/DKFZp762A2316/KIAA1331/SMT3IP2 | |
Rabbit | |
Protein G purification | |
RUO | |
59343 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
SENP2 | |
SENP2 | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human SENP2. | |
400 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only