Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERPINA7 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native SERPINA7.
Supplier: Abnova Corporation PAB5027
Specifications
SERPINA7 | |
Polyclonal | |
Rabbit polyclonal antibody raised against native SERPINA7. | |
In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal) | |
TBG | |
Rabbit | |
150 μg | |
Primary | |
Human | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
SERPINA7 | |
SERPINA7 | |
Native purified human SERPINA7. | |
RUO | |
6906 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction