Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB12, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114853 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-853 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-853 Supplier Abnova Corporation Supplier No. PAB1048

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit polyclonal antibody raised against synthetic peptide of SERPINB12.

Sequence: SHSKDNLKGLEELERK

Specifications

Antigen SERPINB12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description serpin peptidase inhibitor, clade B (ovalbumin), member 12
Formulation In PBS (0.02% sodium azide)
Gene SERPINB12
Gene Alias MGC119247, MGC119248, YUKOPIN
Gene Symbols SERPINB12
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 260-275 of human SERPINB12.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89777
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.