Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERPINB13, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SERPINB13.
Supplier: Abnova Corporation PAB1049
Description
Sequence: SEHKADYSGMSSGSGSpecifications
SERPINB13 | |
Polyclonal | |
serpin peptidase inhibitor, clade B (ovalbumin), member 13 | |
SERPINB13 | |
SERPINB13 | |
A synthetic peptide corresponding to amino acids 310-324 of human SERPINB13. | |
RUO | |
5275 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
In PBS (0.02% sodium azide) | |
HSHUR7SEQ, HUR7, MGC126870, PI13, headpin | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction