Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB14 (OVAL), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89129570 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-570 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-570 Supplier Abnova Corporation Supplier No. PAB28060
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native OVAL.

Specifications

Antigen SERPINB14 (OVAL)
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native OVAL.
Dilution ELISA (1:25000) Immunoprecipitation (1:100) Western Blot (1:500-1:2000) Immunohistochemistry (1:300-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene SERPINB14
Gene Symbols SERPINB14
Host Species Rabbit
Immunogen Native purified OVAL from hen egg white.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 396058
Target Species Chicken
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.