Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINE1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89117339 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-339 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-339 Supplier Abnova Corporation Supplier No. PAB14897
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native SERPINE1.

This gene encodes a member of the serine proteinase inhibitor (serpin) superfamily. This member is the principal inhibitor of tissue plasminogen activator (tPA) and urokinase (uPA), and hence is an inhibitor of fibrinolysis. Defects in this gene are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency), and high concentrations of the gene product are associated with thrombophilia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen SERPINE1
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Inhibition Assays, Radioimmune Assays (RIA), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native SERPINE1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In serum
Gene SERPINE1
Gene Alias PAI/PAI-1/PAI1/PLANH1
Gene Symbols SERPINE1
Host Species Rabbit
Immunogen Native purified human SERPINE1 from the HT 1080 fibrosarcoma cell line.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5054
Test Specificity This antibody reacts with human PAI-1s from endothelial cells. No reactivity with human PAI-2. Identity established by double diffusion with human PAI-1 isolated from human endothelial cells (HUVEC) culture and recombinant PAI-1 expressed in E.coli. No other proteins so far tested showed greater than 1% cross reaction.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.