Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-SERPINF2, Polyclonal, Abnova™

Catalog No. 89116864 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-864 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-864 Supplier Abnova Corporation Supplier No. PAB13872

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Antigen SERPINF2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In 50mM sodium phosphate buffer, 100mM NaCl, 1mM EDTA, pH 6.6
Gene Symbols Serpinf2
Host Species Rabbit
Immunogen Recombinant protein corresponding to mouse Serpinf2.
Quantity 1 mg
Primary or Secondary Primary
Gene ID (Entrez) 18816
Target Species Mouse
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.