Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

serum, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89138024 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-024 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-024 Supplier Abnova Corporation Supplier No. PAB14899
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against serum.

Specifications

Antigen serum
Applications ELISA, Immunodiffusion, Immunomicroscopy
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against serum.
Dilution ELISA (1:1000) Immunodiffusion (1:32) The optimal working dilution should be determined by the end user.
Formulation In serum
Host Species Rabbit
Immunogen Native from whole human serum.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.