Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SETD1A Rabbit anti-Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89116919 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-919 Supplier Abnova Corporation Supplier No. PAB14083
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Setd1a.

Specifications

Antigen SETD1A
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Setd1a.
Dilution ELISA (1:100) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In serum (0.05% sodium azide)
Gene Setd1a
Gene Alias BC010250/KMT2F/MGC55143/Nsccn1/mKIAA0339/mNSC1
Gene Symbols Setd1a
Host Species Rabbit
Immunogen A mixture of synthetic peptides (conjugated with KLH), of which two correspond to N-terminus and one to internal region of mouse Setd1a.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233904
Target Species Mouse
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.