Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SETD1B Rabbit anti-Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89116920 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-920 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-920 Supplier Abnova Corporation Supplier No. PAB14084
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Setd1b.

Specifications

Antigen SETD1B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Setd1b.
Dilution ELISA (1:100-1:500) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In serum (0.05% sodium azide)
Gene Setd1b
Gene Alias AA516740/BC035291/KMT2G/MGC39035/mKIAA1076
Gene Symbols Setd1b
Host Species Rabbit
Immunogen A mixture of synthetic peptides (conjugated with KLH) that corresponding to internal region of mouse Setd1b.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 208043
Target Species Mouse
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.