Learn More
SFRP5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SFRP5.
Supplier: Abnova Corporation PAB0957
Description
Secreted frizzled-related protein 5 (SFRP5) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. SFRP5 and SFRP1 may be involved in determining the polarity of photoreceptor cells in the retina. SFRP5 is highly expressed in the retinal pigment epithelium, and moderately expressed in the pancreas. [provided by RefSeq
Sequence: APARCEEYDYYGWQSpecifications
SFRP5 | |
Polyclonal | |
secreted frizzled-related protein 5 | |
SFRP5 | |
SFRP5 | |
A synthetic peptide corresponding to amino acids 25-38 of human SFRP5. | |
RUO | |
6425 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
In PBS (0.02% sodium azide) | |
SARP3 | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only