Learn More
SFRS11, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SFRS11.
Supplier: Abnova Corporation PAB20446
Description
This gene encodes 54kDa nuclear protein that contains an arginine/serine-rich region similar to segments found in pre-mRNA splicing factors. Although the function of this protein is not yet known, structure and immunolocalization data suggest that it may play a role in pre-mRNA processing. [provided by RefSeq
Sequence: LLPTPNPLTQIGAVPLAALGAPTLDPALAALGLPGANLNSQSLAADQLLKLMSTVDPKLNHVAAGLVSPSLKSDTSSKEIEEAMKRVREAQSLISAAIEPDKKEEKRRHSRSRSRSpecifications
SFRS11 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant SFRS11. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
DKFZp686M13204/dJ677H15.2/p54 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
9295 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
SFRS11 | |
SFRS11 | |
Recombinant protein corresponding to amino acids of human SFRS11. | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only