Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFTSV HB29 Rabbit anti-Viruses, Polyclonal Antibody, Abnova™

Catalog No. 89138420 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-420 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-420 Supplier Abnova Corporation Supplier No. PAB27174
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SFTSV HB29.

Specifications

Antigen SFTSV HB29
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SFTSV HB29.
Formulation In PBS (0.02% sodium azide)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acids at C-terminus of human SFTSV HB29.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity SFTSV HB29 Membrane Glycoprotein antibody is specific to the SFTSV HB29.
Target Species Virus
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.