Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGMS1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89114820 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-820 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-820 Supplier Abnova Corporation Supplier No. PAB10379

Rabbit polyclonal antibody raised against full-length recombinant SGMS1.

The protein encoded by this gene is predicted to be a five-pass transmembrane protein. This gene may be predominately expressed in brain. [provided by RefSeq

Specifications

Antigen SGMS1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant SGMS1.
Dilution Western blot (5 to 10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene SGMS1
Gene Alias MGC17342/MOB/MOB1/SMS1/TMEM23
Gene Symbols SGMS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length SGMS1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259230
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.