Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH2B3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121802 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-802 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-802 Supplier Abnova Corporation Supplier No. PAB20298
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant SH2B3.

T-cell activation requires stimulation of the T-cell receptor (TCR; see MIM 186880)-CD3 (see CD3Z; MIM 186780) complex, followed by recruitment of an array of intracellular signaling proteins (e.g., GRB2 (MIM 108355) and PLCG1 (MIM 172420)). Mediating the interaction between the extracellular receptors and intracellular signaling pathways are adaptor proteins such as LAT (MIM 602354), TRIM (MIM 604962), and LNK.[supplied by OMIM

Sequence: FDPPKSSRPKLQAACSSIQEVRRCTRLEMPDNLYTFVLKVKDRTDIIFEVGDEQQLNSWMAELSECTGRGLESTEAEMHIPSALEPSTSSSPRGSTDSLNQGASPGGLLDPACQKTDHFLSCYPWFH

Specifications

Antigen SH2B3
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant SH2B3.
Dilution Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene SH2B3
Gene Alias CELIAC13/LNK
Gene Symbols SH2B3
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human SH2B3.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10019
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.