Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHANK3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89127107 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-107 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-107 Supplier Abnova Corporation Supplier No. PAB25490
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SHANK3.

This gene is a member of the Shank gene family. Shank proteins are multidomain scaffold proteins of the postsynaptic density that connect neurotransmitter receptors, ion channels, and other membrane proteins to the actin cytoskeleton and G-protein-coupled signaling pathways. Shank proteins also play a role in synapse formation and dendritic spine maturation. Mutations in this gene are a cause of autism spectrum disorder (ASD) which is characterized by impairments in social interaction and communication, and restricted behavioral patterns and interests. Mutations in this gene are a major causitive factor in the neurological symptoms of 22q13.3 deletion syndrome. Additional isoforms have been described for this gene but they have not yet been experimentally verified. [provided by RefSeq

Specifications

Antigen SHANK3
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SHANK3.
Dilution Western Blot (1 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene SHANK3
Gene Alias DEL22q13.3/KIAA1650/PROSAP2/PSAP2/SPANK-2
Gene Symbols SHANK3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region of human SHANK3.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85358
Test Specificity At least three alternatively spliced transcript isoforms of SHANK3 are known to exist.
Target Species Human, Mouse
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.