Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHROOM1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89127286 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-286 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-286 Supplier Abnova Corporation Supplier No. PAB25667
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SHROOM1.

SHROOM family members play diverse roles in the development of the nervous system and other tissues (Hagens et al., 2006 [PubMed 16615870]).[supplied by OMIM

Specifications

Antigen SHROOM1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SHROOM1.
Dilution Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene SHROOM1
Gene Alias APXL2/KIAA1960
Gene Symbols SHROOM1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 16 amino acids at C-terminus of human SHROOM1.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134549
Test Specificity SHROOM1 antibody is human, mouse and rat reactive. At least three isoforms of SHROOM1 are known to exist; this antibody will detect only the two largest isoforms. SHROOM1 antibody is predicted to not cross-react with other SHROOM family members.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.