Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIGLEC1, Rabbit, Clone: SP216, Abnova™

Catalog No. 89110322 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-322 Supplier Abnova Corporation Supplier No. MAB12181

Rabbit monoclonal antibody raised against synthetic peptide of SIGLEC1.

This gene encodes a member of the immunoglobulin superfamily. The encoded protein is a lectin-like adhesion molecule that binds glycoconjugate ligands on cell surfaces in a sialic acid-dependent manner. It is a type I transmembrane protein expressed only by a subpopulation of macrophages and is involved in mediating cell-cell interactions. Alternative splicing produces a transcript variant encoding an isoform that is soluble rather than membrane-bound; however, the full-length nature of this variant has not been determined. [provided by RefSeq

Specifications

Antigen SIGLEC1
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone SP216
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of SIGLEC1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene SIGLEC1
Gene Alias CD169/DKFZp667F058/FLJ00051/FLJ00055/FLJ00073/FLJ00411/FLJ32150/SIGLEC-1/SN/dJ1009E24.1
Gene Symbols SIGLEC1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region of human SIGLEC1.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6614
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.