Learn More
SIGLEC10, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SIGLEC10.
Supplier: Abnova Corporation PAB3644
Description
SIGLECs are members of the immunoglobulin superfamily that are expressed on the cell surface. Most SIGLECs have 1 or more cytoplasmic immune receptor tyrosine-based inhibitory motifs, or ITIMs. SIGLECs are typically expressed on cells of the innate immune system, with the exception of the B-cell expressed SIGLEC6 (MIM 604405).[supplied by OMIM
Specifications
SIGLEC10 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of SIGLEC10. | |
In PBS (0.09% sodium azide) | |
MGC126774/PRO940/SIGLEC-10/SLG2 | |
Rabbit | |
Protein A purification | |
RUO | |
89790 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
SIGLEC10 | |
SIGLEC10 | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human SIGLEC10. | |
400 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only