Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIGLEC11 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89118580 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-580 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-580 Supplier Abnova Corporation Supplier No. PAB16743
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SIGLEC11.

SIGLECs are a family of cell surface lectins defined by shared structural motifs in the first 2 immunoglobulin (Ig)-like domains and by their ability to recognize sialic acids via the first Ig V set domain.[supplied by OMIM

Specifications

Antigen SIGLEC11
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SIGLEC11.
Dilution Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene SIGLEC11
Gene Symbols SIGLEC11
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus 18 amino acids of human SIGLEC11.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114132
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.