Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRT3 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89134086 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-086 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-086 Supplier Abnova Corporation Supplier No. PAB9004
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Sirt3.

Sequence: RDLVGPFVLSPRRKDVVQL

Specifications

Antigen SIRT3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Sirt3.
Dilution Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene Sirt3
Gene Alias 2310003L23Rik/AI848213/Sir2l3
Gene Symbols Sirt3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 203-221 of mouse Sirt3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64384
Target Species Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.