Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slc12a1, Rabbit, ATTO390, Polyclonal Antibody, Abnova™

Catalog No. 89130550 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-550 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-550 Supplier Abnova Corporation Supplier No. PAB29037
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Slc12a1.

Specifications

Antigen Slc12a1
Applications Western Blot
Classification Polyclonal
Conjugate Atto 390
Description Rabbit polyclonal antibody raised against synthetic peptide of Slc12a1.
Dilution Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Slc12a1
Gene Alias MGC93479/Nkcc2
Gene Symbols Slc12a1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 33-55 at N-terminus of rat Slc12a1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25065
Test Specificity Detects ∼160 kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.