Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC12A3, Rabbit, HRP, Polyclonal Antibody, Abnova™

Catalog No. 89130281 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-281 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-281 Supplier Abnova Corporation Supplier No. PAB28761
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant Human, Rat SLC12A3.

Specifications

Antigen SLC12A3
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate HRP
Description Rabbit polyclonal antibody raised against recombinant Human, Rat SLC12A3.
Dilution Immunofluorescence Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation PBS, 50% glycerol and 0.09% sodium azide
Gene Alias FLJ96318/NCCT/TSC
Gene Symbols SLC12A3
Host Species Rabbit
Immunogen Recombinant protein corresponding to SLC12A3 N-terminus (amino acids 74-95).
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6559/54300
Test Specificity 1 ug/mL of PAB28761 was sufficient for detection of SLC12A3 ,∼160kDa, in 10ug of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.