Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slc14a2, Rabbit, ATTO488, Polyclonal Antibody, Abnova™

Catalog No. 89130345 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-345 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-345 Supplier Abnova Corporation Supplier No. PAB28822
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant Rat Slc14a2.

Specifications

Antigen Slc14a2
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Atto 488
Description Rabbit polyclonal antibody raised against recombinant Rat Slc14a2.
Dilution Immunofluorescence Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation PBS, 50% glycerol and 0.09% sodium azide
Gene Slc14a2
Gene Alias Slc14a2T/UrT1-C/UrT1-D/testSymbol
Gene Symbols Slc14a2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Slc14a2 C-terminus (amino acids 911-929).
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54302
Test Specificity 1 ug/mL of PAB28822 was sufficient for detection of Slc14a2, ∼97 and 127kDa, in 20ug of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody.
Target Species Mouse, Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.