Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC16A8, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114654 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-654 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-654 Supplier Abnova Corporation Supplier No. PAB10133
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SLC16A8.

Lactate transport across cell membranes is mediated by proton-coupled monocarboxylate transporters, such as SLC16A8 Yoon et al. (1999) [PubMed 10493836].[supplied by OMIM

Specifications

Antigen SLC16A8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SLC16A8.
Dilution Western blot (1 to 5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene SLC16A8
Gene Alias MCT3/REMP
Gene Symbols SLC16A8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to SLC16A8.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23539
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.