Learn More
Description
The vesicular monoamine transporter acts to accumulate cytosolic monoamines into synaptic vesicles, using the proton gradient maintained across the synaptic vesicular membrane. Its proper function is essential to the correct activity of the monoaminergic systems that have been implicated in several human neuropsychiatric disorders. The transporter is a site of action of important drugs, including reserpine and tetrabenazine (Peter et al., 1993 [PubMed 7905859]). See also SLC18A1 (MIM 193002).[supplied by OMIM
Specifications
Specifications
| Antigen | SLC18A2 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | solute carrier family 18 (vesicular monoamine), member 2 |
| Formulation | In PBS (0.02% sodium azide) |
| Gene | SLC18A2 |
| Gene Alias | MGC120477, MGC120478, MGC26538, SVAT, SVMT, VAT2, VMAT2 |
| Gene Symbols | SLC18A2 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.