Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC2A1, Rabbit, Clone: SP168, Abnova™

Catalog No. 89109384 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-384 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-384 Supplier Abnova Corporation Supplier No. MAB11214

Rabbit monoclonal antibody raised against synthetic peptide of SLC2A1.

This gene encodes a major glucose transporter in the mammalian blood-brain barrier. Mutations in this gene have been found in a family with paroxysmal exertion-induced dyskinesia. [provided by RefSeq

Specifications

Antigen SLC2A1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone SP168
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of SLC2A1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene SLC2A1
Gene Alias DYT17/DYT18/GLUT/GLUT1/MGC141895/MGC141896/PED
Gene Symbols SLC2A1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus region of human SLC2A1.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6513
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.