Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC30A10, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89122486 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-122-486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-122-486 Supplier Abnova Corporation Supplier No. PAB20991

Rabbit polyclonal antibody raised against recombinant SLC30A10.

Zinc functions as a cofactor for numerous enzymes, nuclear factors, and hormones and as an intra- and intercellular signal ion. Members of the zinc transporter (ZNT)/SLC30 subfamily of the cation diffusion facilitator family, such as SLC30A10, permit cellular efflux of zinc (Seve et al., 2004 [PubMed 15154973]).[supplied by OMIM

Sequence: GVNMEELMSKLSAVPGISSVHEVHIWELVSGKIIATLHIKYPKDRGYQDASTKIREIFHHAGIHNVTIQFENVDLKEPLEQKDLLLLCNSPCISKGCAKQLCCPPGALPLAHVNGCAEHNGGPSLDTYGSDGLSRRDAREVAIEVS

Specifications

Antigen SLC30A10
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant SLC30A10.
Dilution Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene SLC30A10
Gene Alias DKFZp547M236/ZNT10/ZNT8/ZRC1/ZnT-10
Gene Symbols SLC30A10
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human SLC30A10.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55532
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.