Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC31A1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117142 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-142 Supplier Abnova Corporation Supplier No. PAB14558
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SLC31A1.

Copper is an element essential for life, but excessive copper can be toxic or even lethal to the cell. Therefore, cells have developed sophisticated ways to maintain a critical copper balance, with the intake, export, and intracellular compartmentalization or buffering of copper strictly regulated. The 2 related genes ATP7A (MIM 300011) and ATP7B (MIM 606882), responsible for the human diseases Menkes syndrome (MIM 309400) and Wilson disease (MIM 277900), respectively, are involved in copper export. In S. cerevisiae, the copper uptake genes CTR1, CTR2, and CTR3 have been identified, and in human the CTR1 and CTR2 (MIM 603088) genes have been identified.[supplied by OMIM

Specifications

Antigen SLC31A1
Applications Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SLC31A1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:250) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Tris-citrate/phosphate buffer, pH 7-8 (0.09% sodium azide)
Gene SLC31A1
Gene Alias COPT1/CTR1/MGC75487/hCTR1
Gene Symbols SLC31A1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human SLC31A1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1317
Target Species Human, Mouse, Pig, Rat, Xenopus species, Zebrafish
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.