Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slc35e4, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89134073 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-073 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-073 Supplier Abnova Corporation Supplier No. PAB8988
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Slc35e4.

Sequence: LGAACSLAGELRAPPAGCGFLLVA

Specifications

Antigen Slc35e4
Applications Dot Blot, ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Slc35e4.
Dilution ELISA (1:10000) Dot Blot (1:10000) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene Slc35e4
Gene Alias A330108F03Rik/AI414849/AI536442/MGC28785
Gene Symbols Slc35e4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 172-195 of mouse Slc35e4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103710
Target Species Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.