Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC39A4 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89117998 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-998 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-998 Supplier Abnova Corporation Supplier No. PAB15987
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SLC39A4.

This gene encodes a member of the zinc/iron-regulated transporter-like protein (ZIP) family. The transmembrane protein is required for zinc uptake in the intestine. Mutations in this gene result in acrodermatitis enteropathica, a rare inherited defect in the absorption of dietary zinc. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen SLC39A4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SLC39A4.
Dilution ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Gene SLC39A4
Gene Alias AEZ/FLJ20327/MGC74741/ZIP4
Gene Symbols SLC39A4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human SLC39A4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55630
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.