Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC40A1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89117622 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-622 Supplier Abnova Corporation Supplier No. PAB15509
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SLC40A1.

The protein encoded by this gene is a cell membrane protein that may be involved in iron export from duodenal epithelial cells. Defects in this gene are a cause of hemochromatosis type 4 (HFE4). [provided by RefSeq

Specifications

Antigen SLC40A1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SLC40A1.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl, pH 7.5 (0.05% sodium azide)
Gene SLC40A1
Gene Alias FPN1/HFE4/IREG1/MST079/MSTP079/MTP1/SLC11A3
Gene Symbols SLC40A1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 250-300 of human SLC40A1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30061
Test Specificity Western blot where a band is seen at ∼62 KDa.
Target Species Human, Mouse
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.