Learn More
SLC40A1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SLC40A1.
Supplier: Abnova Corporation PAB5460
Description
The protein encoded by this gene is a cell membrane protein that may be involved in iron export from duodenal epithelial cells. Defects in this gene are a cause of hemochromatosis type 4 (HFE4). [provided by RefSeq
Specifications
SLC40A1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of SLC40A1. | |
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
FPN1/HFE4/IREG1/MST079/MSTP079/MTP1/SLC11A3 | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
SLC40A1 | |
SLC40A1 | |
A synthetic peptide corresponding to N-terminus of human SLC40A1. | |
RUO | |
30061 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only