Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC7A11 Rabbit anti-Human, Mouse, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89117353 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-353 Supplier Abnova Corporation Supplier No. PAB14931
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SLC7A11.

SLC7A11 is a member of a heteromeric Na(+)-independent anionic amino acid transport system highly specific for cystine and glutamate. In this system, designated system Xc(-), the anionic form of cystine is transported in exchange for glutamate.[supplied by OMIM

Specifications

Antigen SLC7A11
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Biotin
Description Rabbit polyclonal antibody raised against synthetic peptide of SLC7A11.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene SLC7A11
Gene Alias CCBR1/xCT
Gene Symbols SLC7A11
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1-50 of human SLC7A11.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23657
Test Specificity This antibody reacts with the human and mouse proteins. Other species have not been tested. This is specific to XCT. By Western blot a band at ∼55 KDa is seen. Immunoprecipitation is not recommended.
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.