Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC7A11 Rabbit anti-Human, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89117361 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-361 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-361 Supplier Abnova Corporation Supplier No. PAB14941
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Slc7a11.

Specifications

Antigen SLC7A11
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Description Rabbit polyclonal antibody raised against synthetic peptide of Slc7a11.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene Slc7a11
Gene Alias 9930009M05Rik/AI451155/sut/xCT
Gene Symbols Slc7a11
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1-50 of mouse Slc7a11.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26570
Test Specificity This antibody reacts with the human protein. It does not react with murine stomach tissue in Western blot. Other species have not been tested. This is specific to XCT. By Western blot a band at ∼55 KDa is seen. No other applications have been tested. Recommended Starting Dilutions: Western Blot: 1 ug/ml (ECL) Immunoprecipitation: Not tested Immunohisto/cytochemistry: Not tested *Optimal working dilutions should be determined by the investigator.
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.