Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC9A3, Rabbit, FITC, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Slc9a3.
Supplier: Abnova Corporation PAB29030
Specifications
SLC9A3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Slc9a3. | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
Nhe3 | |
Rabbit | |
Affinity purification | |
RUO | |
24784 | |
Rat | |
Liquid |
Immunocytochemistry, Western Blot | |
FITC | |
Immunocytochemistry Western blot (1:1000) The optimal working dilution should be determined by the end user. | |
Slc9a3 | |
Slc9a3 | |
A synthetic peptide corresponding to amino acids 809-831 of rat Slc9a3. | |
100 μg | |
Primary | |
Detects ∼84 kDa. | |
Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction