Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLIRP, Rabbit, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89117509 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-509 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-509 Supplier Abnova Corporation Supplier No. PAB15214
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SLIRP.

Steroid receptor RNA activator (SRA, or SRA1; MIM 603819) is a complex RNA molecule containing multiple stable stem-loop structures that functions in coactivation of nuclear receptors. SLIRP interacts with stem-loop structure-7 of SRA (STR7) and modulates nuclear receptor transactivation (Hatchell et al., 2006 [PubMed 16762838]).[supplied by OMIM

Specifications

Antigen SLIRP
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Description Rabbit polyclonal antibody raised against synthetic peptide of SLIRP.
Dilution Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene SLIRP
Gene Alias DC23/C14orf156/DC50/PD04872
Gene Symbols SLIRP
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human SLIRP.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81892
Test Specificity Does not react with mouse. This is specific to SLIRP protein. This antibody may be used for Western blot analysis where a band at ∼12 KDa is be seen, representing the SLIRP protein. May see additional bands above 45 KDa.
Target Species Human, Primate
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.