Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMAD1/SMAD2/SMAD3/SMAD5, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89138328 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-328 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-328 Supplier Abnova Corporation Supplier No. PAB17216
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SMAD1/SMAD2/SMAD3/SMAD5.

Sequence: GPLQWLDKVLTQMGS

Specifications

Antigen SMAD1/SMAD2/SMAD3/SMAD5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SMAD1/SMAD2/SMAD3/SMAD5.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal)
Gene Alias BSP1/JV4-1/JV41/MADH1/MADR1
Gene Symbols SMAD1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 442-456 at C-terminus of human SMAD1/SMAD2/SMAD3/SMAD5.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4086/4087/4088/4090
Target Species Human, Rat
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.