Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMAD3 Rabbit anti-Bovine, Chicken, Clawed frog, Frog, Human, Mouse, Pig, Rat, Zebra fish, Polyclonal Antibody, Abnova™

Catalog No. 89114971 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-971 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-971 Supplier Abnova Corporation Supplier No. PAB11263
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SMAD3.

The protein encoded by this gene belongs to the SMAD, a family of proteins similar to the gene products of the Drosophila gene 'mothers against decapentaplegic' (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein functions as a transcriptional modulator activated by transforming growth factor-beta and is thought to play a role in the regulation of carcinogenesis. [provided by RefSeq]

Specifications

Antigen SMAD3
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SMAD3.
Dilution ELISA (1:10000-1:40000) Western Blot (1:500-1:3000) Immunohistochemistry (1:500-1:3000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene SMAD3
Gene Alias DKFZp586N0721/DKFZp686J10186/HSPC193/HsT17436/JV15-2/MADH3/MGC60396
Gene Symbols SMAD3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 417-425 of human SMAD3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4088
Test Specificity ELISA and western blot show equivalent reactivity against phosphorylated and non-phosphorylated human Smad3. Expect reactivity against Smad1 and Smad5. Negligible reactivity is seen against other Smad family members.
Target Species Bovine, Chicken, Frog, Human, Mouse, Pig, Rat, Zebrafish
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.