Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-SMARCD3, Polyclonal,

Catalog No. ABE1313MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1313MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1313MI Supplier MilliporeSigma™ Supplier No. ABE1313
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects SMARCD3 in Human, Mouse samples, and it is validated for Immunocytochemistry, Western Blotting

SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 3 (UniProt Q6STE5; also known as 60kDa BRG-1/Brm-associated factor subunit C, BAF60C, BRG1-associated factor 60C, Chromatin remodeling complex BAF60C subunit, Mammalian chromatin remodeling complex BRG1-associated factor 60C, SWI/SNF complex 60kDa subunit C, Swp73-like protein) is a nucleus protein encoded by the SMARCD3 or BAF60C gene (Entrez Gene ID 6604) in human. SMACD3/BAF60C protein belongs to the SWI/SNF family of helicases and ATPases that regulate transcriptions by modifying the chromatin structure around the target genes. Anti-SMARCD3 is an immunogen affinity purified rabbit polyclonal antibody with confirmed reactivity toward human and murine species. The immunogen peptide sequence is present in the C-terminal half of both isoform 1 (BAF60C2) and isoform 2 (BAF60C1), and is 100% conserved among human, murine, rat, feline, equine, and bovine species.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCD3
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q6STE5
Gene Symbols SMARCD3; BAF60C
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the SWIB domain of human SMARCD3.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003069
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.