Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMTNL2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89123475 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-475 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-475 Supplier Abnova Corporation Supplier No. PAB21774

Rabbit polyclonal antibody raised against recombinant SMTNL2.

Sequence: SSFTWSVPSSGYGAVTASKHSNSPPLVTPPQSPVSPQPPAITQVHRQGERRRELVRSQTLPRTSEAQARKALFEKWEQETAAGKGKGEARARLKRS

Specifications

Antigen SMTNL2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description smoothelin-like 2
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene SMTNL2
Gene Alias FLJ42461, MGC131847, MGC138382
Gene Symbols SMTNL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human SMTNL2.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342527
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.