Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNCAIP Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114366 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-366 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-366 Supplier Abnova Corporation Supplier No. PAB0717
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of SNCAIP.

This gene encodes a protein containing several protein-protein interaction domains, including ankyrin-like repeats, a coiled-coil domain, and an ATP/GTP-binding motif. The encoded protein interacts with alpha-synuclein in neuronal tissue and may play a role in the formation of cytoplasmic inclusions and neurodegeneration. A mutation in this gene has been associated with Parkinson's disease. Alternatively spliced transcript variants encoding different isoforms of this gene have been described, but their full-length nature has yet to be determined. [provided by RefSeq]

Sequence: SLELNGEKDKDKGRTLQRT

Specifications

Antigen SNCAIP
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SNCAIP.
Dilution Immunohistochemistry (1:1000-1:2000) The optimal dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene SNCAIP
Gene Alias MGC39814/SYPH1
Gene Symbols SNCAIP
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 829-847 of human SNCAIP.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9627
Target Species Human, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.