Learn More
SNTB1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SNTB1.
Supplier: Abnova Corporation PAB21733
Description
Dystrophin is a large, rod-like cytoskeletal protein found at the inner surface of muscle fibers. Dystrophin is missing in Duchenne Muscular Dystrophy patients and is present in reduced amounts in Becker Muscular Dystrophy patients. The protein encoded by this gene is a peripheral membrane protein found associated with dystrophin and dystrophin-related proteins. This gene is a member of the syntrophin gene family, which contains at least two other structurally-related genes. [provided by RefSeq
Sequence: QGCHNSAELIAEISTACTYKNQECRLTIHYENGFSITTEPQEGAFPKTIIQSPYEKLKMSSDDGSpecifications
SNTB1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant SNTB1. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
59-DAP/A1B/BSYN2/DAPA1B/FLJ22442/MGC111389/SNT2/SNT2B1/TIP-43 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
6641 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
SNTB1 | |
SNTB1 | |
Recombinant protein corresponding to amino acids of human SNTB1. | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only