Learn More
SNX3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SNX3.
Supplier: Abnova Corporation PAB5584
Description
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like most family members. This protein interacts with phosphatidylinositol-3-phosphate, and is involved in protein trafficking. [provided by RefSeq
Specifications
SNX3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of SNX3. | |
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
Grd19/MCOPS8/MGC17570/SDP3/SNX3A | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
SNX3 | |
SNX3 | |
A synthetic peptide corresponding to N-terminus of human SNX3. | |
RUO | |
8724 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only