Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD1 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114373 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-373 Supplier Abnova Corporation Supplier No. PAB0725
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Sod1.

Sequence: ASGEPVVLSGQIT

Specifications

Antigen SOD1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Sod1.
Dilution Western Blot (1:500-1:1000) The optimal dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Sod1
Gene Alias B430204E11Rik/Cu/Zn-SOD/CuZnSOD/Ipo-1/Ipo1/MGC107553/SODC/Sod-1
Gene Symbols Sod1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated diphtheria toxoid) corresponding to amino acids 24-36 of mouse Sod1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20655
Target Species Mouse, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.