Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD1 Rabbit anti-Bovine, Polyclonal Antibody, Abnova™

Catalog No. 89129597 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-597 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-597 Supplier Abnova Corporation Supplier No. PAB28092
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native SOD1 (Bovine).

Specifications

Antigen SOD1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native SOD1 (Bovine).
Dilution ELISA (1:10000 - 1:50000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene SOD1
Gene Alias SOD1L1
Gene Symbols SOD1
Host Species Rabbit
Immunogen Native purified SOD1 from bovine erythrocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 281495
Target Species Bovine
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.