Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sod2, Rabbit, ATTO390, Polyclonal Antibody, Abnova™

Catalog No. 89130493 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-493 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-493 Supplier Abnova Corporation Supplier No. PAB28983
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against Recombinant of Sod2.

Specifications

Antigen Sod2
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Atto 390
Description Rabbit polyclonal antibody raised against Recombinant of Sod2.
Dilution ELISA Immunofluorescence Immunohistochemistry Immunoprecipitation Western blot (0.5 ug/ml) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Sod2
Gene Symbols Sod2
Host Species Rabbit
Immunogen Recombinant protien corresponding rat Sod2.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24787
Test Specificity Detects a ∼25 kDa protein corresponding to the molecular mass of Mn superoxide dismutase (SOD) on SDS PAGE immunoblots.
Target Species Bovine, Canine, Chicken, Drosophila, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus species
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.